missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-nitroacetophenone 98.0+%, TCI America™

Catalog No. C33081G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33081G Supplier TCI America Supplier No. C33081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23082-51-1
Molecular Formula C8H6ClNO3
Molecular Weight (g/mol) 199.59
MDL Number MFCD09031331
InChI Key PUUYGMZERWRIDS-UHFFFAOYSA-N
PubChem CID 12643796
IUPAC Name 1-(4-chloro-2-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=C(C=C(Cl)C=C1)[N+]([O-])=O

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 157°C
Quantity 1 g
Formula Weight 199.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-2′-nitroacetophenone