accessibility menu, dialog, popup

UserName

CAS RN 23082-51-1

IUPAC Name: 1-(4-chloro-2-nitrophenyl)ethan-1-one
Synonyms: 1-(4-chloro-2-nitrophenyl)ethanone
Molecular Weight (g/mol): 199.59
Molecular Formula: C8H6ClNO3
InChi Key: PUUYGMZERWRIDS-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C=C(Cl)C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results