missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(methylthio)pyrimidine 98.0+%, TCI America™

Catalog No. p-7125820
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30875G 5 g
C308725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30875G Supplier TCI America Supplier No. C30875G

Chemical Identifiers

CAS 49844-90-8
Molecular Formula C5H5ClN2S
Molecular Weight (g/mol) 160.62
MDL Number MFCD00006083
InChI Key DFOHHQRGDOQMKG-UHFFFAOYSA-N
Synonym 4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
PubChem CID 97113
IUPAC Name 4-chloro-2-(methylsulfanyl)pyrimidine
SMILES CSC1=NC=CC(Cl)=N1

Specifications

Boiling Point 112°C
Quantity 5 g
UN Number 1760
Formula Weight 160.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-(methylthio)pyrimidine