missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylquinoline, 97%

Catalog No. p-7022295
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500203 1 g
AAA1500206 5 g
AAA1500214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500203 Supplier Thermo Scientific Chemicals Supplier No. A1500203
Only null left

CAS: 4295-06-1 | C10H8ClN | 177.63 g/mol

4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 4295-06-1
MDL Number MFCD00006757

Specifications

Melting Point ∼25°C
Density 0.881
Boiling Point 269°C to 270°C
Flash Point >110°C (230°F)
Molecular Formula C10H8ClN
Refractive Index 1.622
Quantity 1 g
Beilstein 3522
Synonym 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
Solubility Information Slightly soluble in water.
InChI Key HQAIROMRVBVWSK-UHFFFAOYSA-N
SMILES CC1=NC2=CC=CC=C2C(=C1)Cl
IUPAC Name 4-chloro-2-methylquinoline
Molecular Weight (g/mol) 177.63
PubChem CID 77973
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylquinoline
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-300-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only