accessibility menu, dialog, popup

UserName

H3CClN

CAS RN 4295-06-1

IUPAC Name: 4-chloro-2-methylquinoline
Synonyms: 4-chloro-2-methyl-quinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: HQAIROMRVBVWSK-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C2C=CC=CC2=N1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)