missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylquinoline, 97%

Catalog No. AAA1500206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500206 5 g
AAA1500203 1 g
AAA1500214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500206 Supplier Thermo Scientific Chemicals Supplier No. A1500206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4295-06-1 | C10H8ClN | 177.63 g/mol

4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 4295-06-1
MDL Number MFCD00006757

Specifications

Melting Point ∼25°C
Density 0.881
Boiling Point 269°C to 270°C
Flash Point >110°C (230°F)
Molecular Formula C9H6F3O3
Refractive Index 1.622
Quantity 5 g
Beilstein 3522
Synonym 4-trifluoromethoxy phenylacetic acid, 2-4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenylaceticacid, 4-trifluoromethoxy-phenyl-acetic acid, 4-trifluoromethoxyphenylacetic acid, benzeneacetic acid, 4-trifluoromethoxy, maybridge3_006163
Solubility Information Slightly soluble in water.
InChI Key ZMTIBXQPXBUWPQ-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1CC(=O)O)OC(F)(F)F
IUPAC Name 2-[4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol) 219.14
PubChem CID 2777319
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylquinoline
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-300-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only