missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-methylpyrimidine 98.0+%, TCI America™
$177.08 - $613.27
Chemical Identifiers
| CAS | 4994-86-9 |
|---|---|
| Molecular Formula | C5H5ClN2 |
| Molecular Weight (g/mol) | 128.559 |
| MDL Number | MFCD00234069 |
| InChI Key | WDTVJRYCMIZPMX-UHFFFAOYSA-N |
| PubChem CID | 424447 |
| IUPAC Name | 4-chloro-2-methylpyrimidine |
| SMILES | CC1=NC=CC(=N1)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2835200MG
|
TCI America
C2835200MG |
200 mg |
Each for $177.08
|
|
|||||
|
C28351G
|
TCI America
C28351G |
1 g |
Each for $613.27
|
|
|||||
Chemical Identifiers
| 4994-86-9 | |
| 128.559 | |
| WDTVJRYCMIZPMX-UHFFFAOYSA-N | |
| 4-chloro-2-methylpyrimidine |
| C5H5ClN2 | |
| MFCD00234069 | |
| 424447 | |
| CC1=NC=CC(=N1)Cl |
Specifications
| 4994-86-9 | |
| White-Yellow | |
| C5H5ClN2 | |
| 200 mg | |
| CC1=NC=CC(=N1)Cl | |
| 128.559 | |
| 128.56 | |
| Crystalline Powder |
| 57°C | |
| 168°C | |
| MFCD00234069 | |
| WDTVJRYCMIZPMX-UHFFFAOYSA-N | |
| 4-chloro-2-methylpyrimidine | |
| 424447 | |
| ≥98.0% (GC) | |
| 4-Chloro-2-methylpyrimidine |
Safety and Handling
TSCA : No