CAS RN 4994-86-9
IUPAC Name:
4-chloro-2-methylpyrimidine
Synonyms:
4-chloro-2-methylpyrimidine
Molecular Weight (g/mol):
128.56
Molecular Formula:
C5H5ClN2
InChi Key:
WDTVJRYCMIZPMX-UHFFFAOYSA-N
SMILES:
CC1=NC=CC(Cl)=N1
1
–
1
of
1
results
1
4-Chloro-2-methylpyrimidine 98.0+%, TCI America™
CAS: 4994-86-9 Molecular Formula: C5H5ClN2 Molecular Weight (g/mol): 128.559 MDL Number: MFCD00234069 InChI Key: WDTVJRYCMIZPMX-UHFFFAOYSA-N PubChem CID: 424447 IUPAC Name: 4-chloro-2-methylpyrimidine SMILES: CC1=NC=CC(=N1)Cl