missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylpyrimidine 98.0+%, TCI America™

Catalog No. C2835200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2835200MG 200 mg
C28351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2835200MG Supplier TCI America Supplier No. C2835200MG

Chemical Identifiers

CAS 4994-86-9
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00234069
InChI Key WDTVJRYCMIZPMX-UHFFFAOYSA-N
PubChem CID 424447
IUPAC Name 4-chloro-2-methylpyrimidine
SMILES CC1=NC=CC(=N1)Cl

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 168°C
Quantity 200 mg
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-methylpyrimidine