missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. C30691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30691G 1 g
C30695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30691G Supplier TCI America Supplier No. C30691G
Only null left

Chemical Identifiers

CAS 129716-11-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD06201140
InChI Key AHXBDGJNZJKLNE-UHFFFAOYSA-N
Synonym 4-chloro-2-methylphenyl methanol, 4-chloro-2-methylbenzyl alcohol, benzenemethanol, 4-chloro-2-methyl, acmc-20aaxd, 4-chloro-2-methylbenzylalcohol, 4-chloro-2-methyl-phenyl-methanol
PubChem CID 2757669
IUPAC Name (4-chloro-2-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)Cl)CO

Specifications

Melting Point 62°C
Color White
Quantity 1 g
Formula Weight 156.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-methylbenzyl Alcohol