missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. C25085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25085G 5 g
C250825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25085G Supplier TCI America Supplier No. C25085G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7499-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045852
InChI Key XXFKOBGFMUIWDH-UHFFFAOYSA-N
Synonym 4-chloro-2-methylbenzoicacid, 4-chloro-o-toluic acid, benzoic acid, 4-chloro-2-methyl, 2-methyl-4-chlorobenzoic acid, 4-chloro-2-methyl-benzoic acid, pubchem4570, acmc-1bjnb, o-toluic acid, 4-chloro, ksc497q4r, 4-chloro-2-methylbenzoic acid
PubChem CID 348269
IUPAC Name 4-chloro-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Cl)C(=O)O

Specifications

Melting Point 173°C
Quantity 5 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-methylbenzoic Acid