missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylbenzoic acid, 98%

Catalog No. p-7029762
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2726203 1 g
AAH2726206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2726203 Supplier Thermo Scientific Chemicals Supplier No. H2726203
Only null left

CAS: 7499-07-2 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7499-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045852

Specifications

Melting Point 170°C to 174°C
Quantity 1 g
Synonym 3-chloro-o-toluic acid, 3-chloro-2-methyl-benzoic acid, benzoic acid, 3-chloro-2-methyl, o-toluic acid, 3-chloro, 3-chloro-2-methylbenzoicacid, 2-methyl-3-chlorobenzoic acid, pubchem8362, 2-carboxy-6-chlorotoluene, 3chloro-2-methylbenzoic acid, 3-chloro-2-methybenzoic acid
InChI Key HXGHMCLCSPQMOR-UHFFFAOYSA-N
SMILES CC1=C(C=CC=C1Cl)C(=O)O
IUPAC Name 3-chloro-2-methylbenzoic acid
PubChem CID 82010
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methylbenzoic acid
Hazard Category H302-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only