missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylaniline 98.0+%, TCI America™

Catalog No. p-7117016
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A070425G 25 g
A0704100G 100 g
A0704500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A070425G Supplier TCI America Supplier No. A070425G

Chemical Identifiers

CAS 95-69-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007842
InChI Key CXNVOWPRHWWCQR-UHFFFAOYSA-N
Synonym 4-chloro-o-toluidine, 2-amino-5-chlorotoluene, fast red tr base, p-chloro-o-toluidine, kambamine red tr, benzenamine, 4-chloro-2-methyl, red tr base, 5-chloro-2-aminotoluene, 2-methyl-4-chloroaniline, 4-chloro-2-toluidine
PubChem CID 7251
ChEBI CHEBI:82276
IUPAC Name 4-chloro-2-methylaniline
SMILES CC1=C(C=CC(=C1)Cl)N

Specifications

Color White-Yellow
Boiling Point 241°C
Quantity 25 g
UN Number 2239
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloro-2-methylaniline
EINECSNumber (3)-0265
RTECSNumber XU5000000
TSCA Yes