missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylaniline, 98%

Catalog No. AC109370050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109370050 5 g Glass bottle
AC109371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109370050 Supplier Thermo Scientific Chemicals Supplier No. 109370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 95-69-2 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-69-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00007842
InChI Key CXNVOWPRHWWCQR-UHFFFAOYSA-N
Synonym 4-chloro-o-toluidine, 2-amino-5-chlorotoluene, fast red tr base, p-chloro-o-toluidine, kambamine red tr, benzenamine, 4-chloro-2-methyl, red tr base, 5-chloro-2-aminotoluene, 2-methyl-4-chloroaniline, 4-chloro-2-toluidine
PubChem CID 7251
ChEBI CHEBI:82276
IUPAC Name 4-chloro-2-methylaniline
SMILES CC1=C(C=CC(=C1)Cl)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Density 1.1900g/mL
Boiling Point 241.0°C
Flash Point 124°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5845 to 1.5865
Linear Formula ClC6H3(CH3)NH2
Quantity 5 g
Beilstein 12,835
Specific Gravity 1.19
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methylaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Toxic if swallowed.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-441-6

RUO – Research Use Only