missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-methylacetoacetanilide 98.0+%, TCI America™

Catalog No. C210425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C210425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C210425G Supplier TCI America Supplier No. C210425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20139-55-3
Molecular Formula C11H12ClNO2
Molecular Weight (g/mol) 225.67
MDL Number MFCD00018493
InChI Key ODFRAIZRJMUPCP-UHFFFAOYSA-N
Synonym N-(4-Chloro-o-tolyl)acetoacetamide, N-Acetoacetyl-4-chloro-o-toluidine, N-Acetoacetyl-4-chloro-2-methylaniline
PubChem CID 258549
IUPAC Name N-(4-chloro-2-methylphenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(Cl)C=C1C

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 25 g
Formula Weight 225.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-2′-methylacetoacetanilide
EINECSNumber (3)-0272
RTECSNumber AK4599000
TSCA Yes