accessibility menu, dialog, popup

UserName

CAS RN 20139-55-3

IUPAC Name: N-(4-chloro-2-methylphenyl)-3-oxobutanamide
Synonyms: N-(4-chloro-2-methylphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 225.67
Molecular Formula: C11H12ClNO2
InChi Key: ODFRAIZRJMUPCP-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=C(Cl)C=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results