missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog No. p-3771079
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC398560050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398560050 Supplier Thermo Scientific Chemicals Supplier No. 398560050

CAS: 90296-27-8 | Molecular Formula: C8H9ClO2 | Molecular Weight: 172.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90296-27-8
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.61
MDL Number MFCD02683547
InChI Key AXXPZAITCCIOIA-UHFFFAOYSA-N
Synonym 4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
PubChem CID 3549393
IUPAC Name (4-chloro-2-methoxyphenyl)methanol
SMILES COC1=C(CO)C=CC(Cl)=C1

Specifications

Melting Point 47°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H3(OH)OCH3
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 172.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methoxybenzyl alcohol
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only