missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-iodobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7125552
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26041G 1 g
C26045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26041G Supplier TCI America Supplier No. C26041G

Chemical Identifiers

CAS 13421-13-1
Molecular Formula C7H4ClIO2
Molecular Weight (g/mol) 282.46
MDL Number MFCD00060676
InChI Key LRRDANNSUCQNDU-UHFFFAOYSA-N
Synonym 4-chloro-2-iodobenzoicacid, 2-iodo-4-chlorobenzoic acid, 4-chloro-2-iodo-benzoic acid, benzoic acid, 4-chloro-2-iodo, buttpark 100\07-49, pubchem4018, acmc-209bu6, ksc519a3p, benzoic acid,4-chloro-2-iodo, 4-chloro-2-iodobenzoic acid
PubChem CID 139452
IUPAC Name 4-chloro-2-iodobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C=C1I

Specifications

Melting Point 164°C
Quantity 1 g
Formula Weight 282.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-iodobenzoic Acid