missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorotoluene 98.0+%, TCI America™

Catalog No. C151325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C151325G 25 g
C1513250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C151325G Supplier TCI America Supplier No. C151325G
Only null left

Chemical Identifiers

CAS 452-75-5
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.57
MDL Number MFCD00000571
InChI Key MKFCYQTVSDCXAQ-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene
PubChem CID 520630
IUPAC Name 4-chloro-2-fluoro-1-methylbenzene
SMILES CC1=CC=C(Cl)C=C1F

Specifications

Color Yellow
Quantity 25 g
UN Number 1993
Formula Weight 144.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluorotoluene