missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoropyridine, 95%

Catalog No. p-7055866
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6455703 1 g
AAH6455706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6455703 Supplier Thermo Scientific Chemicals Supplier No. H6455703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34941-92-9 | C5H3ClFN | 131.534 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34941-92-9
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD04112503
InChI Key GNJKJKBURMCLOR-UHFFFAOYSA-N
Synonym 2-fluoro-4-chloropyridine, 4-chloro-2-fluoro-pyridine, pyridine, 4-chloro-2-fluoro, pubchem13535, pyridine,4-chloro-2-fluoro
PubChem CID 2762839
IUPAC Name 4-chloro-2-fluoropyridine
SMILES C1=CN=C(C=C1Cl)F

Specifications

Quantity 1 g
Beilstein 1634727
Formula Weight 131.53
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-fluoropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only