CAS RN 34941-92-9
IUPAC Name:
4-chloro-2-fluoropyridine
Synonyms:
4-chloro-2-fluoropyridine
Molecular Weight (g/mol):
131.53
Molecular Formula:
C5H3ClFN
InChi Key:
GNJKJKBURMCLOR-UHFFFAOYSA-N
SMILES:
FC1=NC=CC(Cl)=C1
1
–
1
of
1
results
1
Promotions Available
4-Chloro-2-fluoropyridine, 95%
4-Chloro-2-fluoropyridine, 95%
CAS: 34941-92-9 Molecular Formula: C5H3ClFN Molecular Weight (g/mol): 131.534 MDL Number: MFCD04112503 InChI Key: GNJKJKBURMCLOR-UHFFFAOYSA-N Synonym: 2-fluoro-4-chloropyridine,4-chloro-2-fluoro-pyridine,pyridine, 4-chloro-2-fluoro,pubchem13535,pyridine,4-chloro-2-fluoro PubChem CID: 2762839 IUPAC Name: 4-chloro-2-fluoropyridine SMILES: C1=CN=C(C=C1Cl)F