missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C30481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30481G Supplier TCI America Supplier No. C30481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 160591-91-3
Melting Point 250°C
Color White-Yellow
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD02684293
Synonym 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
InChI Key YBNDRTRLXPEWKQ-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)Cl)F)(O)O
IUPAC Name (4-chloro-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 4193595
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less