missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoronitrobenzene 96.0+%, TCI America™

Catalog No. C12615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12615G 5 g
C126125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12615G Supplier TCI America Supplier No. C12615G

Chemical Identifiers

CAS 700-37-8
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.54
MDL Number MFCD00042211
InChI Key PTCPUGKKWNMITF-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoronitrobenzene, 2-fluoro-4-chloronitrobenzene, 1-chloro-3-fluoro-4-nitrobenzene, benzene, 4-chloro-2-fluoro-1-nitro, 4-chloro-2-fluoro-1-nitro-benzene, 4-chlor-2-fluor-1-nitrobenzol, pubchem1051, acmc-1bl8e, 4-chloro2-fluoronitrobenzene, ksc380m1b
PubChem CID 69691
IUPAC Name 4-chloro-2-fluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C=C1F

Specifications

Melting Point 48°C
Quantity 5 g
Formula Weight 175.54
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-fluoronitrobenzene