missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7125487
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24835G 5 g
C248325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24835G Supplier TCI America Supplier No. C24835G

Chemical Identifiers

CAS 71916-82-0
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD00831105
InChI Key UFCSSWZQROEFBZ-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-chloro-2-fluorobenzene, 4-chloro-2-fluorobenzyl bromide, 2-fluoro-4-chlorobenzyl bromide, 4-chloro-2-fluorobenzylbromide, alpha-bromo-4-chloro-2-fluorotoluene, benzene, 1-bromomethyl-4-chloro-2-fluoro, pubchem4897, 1-bromomethyl-4-chloro-2-fluoro-benzene, acmc-209okn
PubChem CID 2736551
IUPAC Name 1-(bromomethyl)-4-chloro-2-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)F)CBr

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
UN Number 3265
Formula Weight 223.47
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluorobenzyl Bromide
TSCA No
Recommended Storage Refrigerator