missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7125293
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21215G 5 g
C212125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21215G Supplier TCI America Supplier No. C21215G
Only null left

Chemical Identifiers

CAS 57381-51-8
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00143284
InChI Key JRDMGVGCATYZPW-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzonitrile, 4-chloro-2-fluoro-benzonitrile, benzonitrile, 4-chloro-2-fluoro, 4-chloro-2-fluorobenzenecarbonitrile, ncr dg bf, pubchem3424, 1-chloro-3-fluorobezene, acmc-1b0zj, ksc493o3h
PubChem CID 93655
IUPAC Name 4-chloro-2-fluorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)F)C#N

Specifications

Melting Point 60°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 155.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chloro-2-fluorobenzonitrile