accessibility menu, dialog, popup

UserName

FClN

CAS RN 57381-51-8

IUPAC Name: 4-chloro-2-fluorobenzonitrile
Synonyms: 4-chloro-2-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: JRDMGVGCATYZPW-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC=C1C#N
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)