missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluorobenzaldehyde, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 61072-56-8 |
|---|---|
| Molecular Formula | C7H4ClFO |
| Molecular Weight (g/mol) | 158.56 |
| InChI Key | UVGYSEIWAOOIJR-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985 |
| PubChem CID | 2724908 |
| IUPAC Name | 4-chloro-2-fluorobenzaldehyde |
| SMILES | C1=CC(=C(C=C1Cl)F)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC290620050
|
Thermo Scientific Chemicals
290620050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC290620250
|
Thermo Scientific Chemicals
290620250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 61072-56-8 | |
| 158.56 | |
| 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985 | |
| 4-chloro-2-fluorobenzaldehyde |
| C7H4ClFO | |
| UVGYSEIWAOOIJR-UHFFFAOYSA-N | |
| 2724908 | |
| C1=CC(=C(C=C1Cl)F)C=O |
Specifications
| 61072-56-8 | |
| 98.5% min. (GC) | |
| C7H4ClFO | |
| 5g | |
| Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents | |
| C1=CC(=C(C=C1Cl)F)C=O | |
| 158.56 | |
| 158.56 | |
| 4-Chloro-2-fluorobenzaldehyde |
| Authentic | |
| Glass bottle | |
| ClC6H3(F)CHO | |
| 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985 | |
| UVGYSEIWAOOIJR-UHFFFAOYSA-N | |
| 4-chloro-2-fluorobenzaldehyde | |
| 2724908 | |
| 99% |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only