missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-6-iodoaniline, 96%

Catalog No. AAL1765803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1765803 1 g
AAL1765806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1765803 Supplier Thermo Scientific Chemicals Supplier No. L1765803

CAS: 216393-67-8 | C6H4ClFIN | 271.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216393-67-8
Molecular Formula C6H4ClFIN
Molecular Weight (g/mol) 271.46
MDL Number MFCD01318122
InChI Key SSNQXCONXNVTJN-UHFFFAOYSA-N
Synonym benzenamine,4-chloro-2-fluoro-6-iodo, pubchem2962, 2-fluoro-4-chloro-6-iodoaniline, 2-iodo-4-chloro-6-fluoroaniline, 4-chloro-2-fluoro-6-iodo-phenylamine, 4-chloranyl-2-fluoranyl-6-iodanyl-aniline
PubChem CID 2773650
IUPAC Name 4-chloro-2-fluoro-6-iodoaniline
SMILES NC1=C(I)C=C(Cl)C=C1F

Specifications

Melting Point 39°C to 41°C
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 271.46
Percent Purity 96%
Chemical Name or Material 4-Chloro-2-fluoro-6-iodoaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only