missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-1-iodobenzene 98.0+%, TCI America™

Catalog No. p-7125018
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14995G 5 g
C149925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14995G Supplier TCI America Supplier No. C14995G
Only null left

Chemical Identifiers

CAS 6797-79-1
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00079729
InChI Key RSTFBOIFYXJIMR-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoroiodobenzene, 2-fluoro-4-chloroiodobenzene, 1-chloro-3-fluoro-4-iodobenzene, 4-chloro-2-fluoro-1-iodo-benzene, 4-chloro-2-fluoro-iodobenzene, pubchem3435, acmc-1b71k, 2-iodo-5-chlorofluorobenzene, 2-iodo-5-chlorofluoro benzene, 3-fluoro-4-iodophenyl chloride
PubChem CID 2736562
IUPAC Name 4-chloro-2-fluoro-1-iodobenzene
SMILES FC1=C(I)C=CC(Cl)=C1

Specifications

Quantity 5 g
Formula Weight 256.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluoro-1-iodobenzene