missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-1-iodobenzene, 98%

Catalog No. AAA1937606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1937606 5 g
AAA1937614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1937606 Supplier Thermo Scientific Chemicals Supplier No. A1937606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6797-79-1 | C6H3ClFI | 256.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6797-79-1
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00079729
InChI Key RSTFBOIFYXJIMR-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoroiodobenzene, 2-fluoro-4-chloroiodobenzene, 1-chloro-3-fluoro-4-iodobenzene, 4-chloro-2-fluoro-1-iodo-benzene, 4-chloro-2-fluoro-iodobenzene, pubchem3435, acmc-1b71k, 2-iodo-5-chlorofluorobenzene, 2-iodo-5-chlorofluoro benzene, 3-fluoro-4-iodophenyl chloride
PubChem CID 2736562
IUPAC Name 4-chloro-2-fluoro-1-iodobenzene
SMILES FC1=C(I)C=CC(Cl)=C1

Specifications

Density 1.98
Boiling Point 217°C
Flash Point >110°C (230°F)
Refractive Index 1.606
Quantity 5 g
Beilstein 6288419
Sensitivity Light sensitive
Formula Weight 256.45
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoro-1-iodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only