missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-dimethylphenol, 98%

Catalog No. AAA1364909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1364909 10 g
AAA1364918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1364909 Supplier Thermo Scientific Chemicals Supplier No. A1364909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1123-63-3 | C8H9ClO | 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1123-63-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00020138
InChI Key VWYKSJIPZHRLNO-UHFFFAOYSA-N
PubChem CID 70747
IUPAC Name 4-chloro-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)Cl

Specifications

Melting Point 80°C to 83°C
Quantity 10 g
Beilstein 1908282
Synonym 4-chloro-2,6-xylenol, phenol, 4-chloro-2,6-dimethyl, 2,6-dimethyl-4-chlorophenol, 4-chloro-2,6-dimethyl-phenol, 2,6-xylenol, 4-chloro, pubchem2657, acmc-1c4bc, phenol,4-chloro-2,6-dimethyl, 4-chloro-2,6-xylenol oh=1, 4-chloro-2,6-dimethylphenol
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,6-dimethylphenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-376-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only