missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-difluorobenzoic acid, 97%

Catalog No. AAH2655603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2655603 1 g
AAH2655606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2655603 Supplier Thermo Scientific Chemicals Supplier No. H2655603

CAS: 196194-58-8 | C7H3ClF2O2 | 192.546 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 196194-58-8
Molecular Formula C7H3ClF2O2
Molecular Weight (g/mol) 192.546
MDL Number MFCD06203570
InChI Key ZCJKTGPZLLGECQ-UHFFFAOYSA-N
PubChem CID 17750684
IUPAC Name 4-chloro-2,6-difluorobenzoic acid
SMILES C1=C(C=C(C(=C1F)C(=O)O)F)Cl

Specifications

Melting Point 183°C to 186°C
Quantity 1 g
Synonym benzoic acid, 4-chloro-2,6-difluoro, acmc-20aglx, 4-chloro-2,6-difluorobenzoicacid, 4-chloro-2,6-difluoro-benzoic acid, benzoic acid,4-chloro-2,6-difluoro, 4-chloranyl-2,6-bis fluoranyl benzoic acid
Formula Weight 192.55
Percent Purity 97%
Chemical Name or Material 4-Chloro-2,6-difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only