missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-dibromoaniline 98.0+%, TCI America™

Catalog No. B151625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B151625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B151625G Supplier TCI America Supplier No. B151625G
Only null left

Chemical Identifiers

CAS 874-17-9
Molecular Formula C6H4Br2ClN
Molecular Weight (g/mol) 285.363
MDL Number MFCD00051750
InChI Key XEYLQXUJSOJWJV-UHFFFAOYSA-N
Synonym 4-chloro-2,6-dibromoaniline, 2,6-dibromo-4-chloro-phenylamine, 4-chloro-4,6-dibromoaniline, 2,6-dibromo-4-chlorophenylamine, pubchem3632, acmc-209qkf, 2,6-dibromo-4-chloro-aniline, zerenex e/9070016, labotest-bb lt01147813, labotest-bb lt03332130
PubChem CID 625286
IUPAC Name 2,6-dibromo-4-chloroaniline
SMILES C1=C(C=C(C(=C1Br)N)Br)Cl

Specifications

Melting Point 93°C
Boiling Point 301°C
Quantity 25 g
Formula Weight 285.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,6-dibromoaniline