missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2,5-dimethylaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 155.63 |
| Percent Purity | ≥98.0% (GC,T) |
| Chemical Name or Material | 4-Chloro-2,5-dimethylaniline |
Safety and Handling
| TSCA | No |