missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2,5-dimethoxyaniline 98.0+%, TCI America™
Supplier: TCI America C199625G
Specifications
| 4-Chloro-2,5-dimethoxyaniline | |
| 119°C | |
| MFCD00014893 | |
| 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718 | |
| COC1=CC(Cl)=C(OC)C=C1N | |
| 187.62 | |
| 187.62 | |
| Crystalline Powder |
| 6358-64-1 | |
| C8H10ClNO2 | |
| 25 g | |
| YGUFQYGSBVXPMC-UHFFFAOYSA-N | |
| 4-chloro-2,5-dimethoxyaniline | |
| 22833 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 6358-64-1 | |
| 187.62 | |
| YGUFQYGSBVXPMC-UHFFFAOYSA-N | |
| 22833 | |
| COC1=CC(Cl)=C(OC)C=C1N |
| C8H10ClNO2 | |
| MFCD00014893 | |
| 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718 | |
| 4-chloro-2,5-dimethoxyaniline |
Safety and Handling
EINECSNumber : (3)-0943
RTECSNumber : BX1225000
TSCA : Yes