missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2',5'-dimethoxyacetoacetanilide 98.0+%, TCI America™

Catalog No. C210025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C210025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C210025G Supplier TCI America Supplier No. C210025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4433-79-8
Molecular Formula C12H14ClNO4
Molecular Weight (g/mol) 271.70
MDL Number MFCD00026256
InChI Key MOUVJGIRLPZEES-UHFFFAOYSA-N
Synonym N-(4-Chloro-2,5-dimethoxyphenyl)acetoacetamide, Naphthol AS-IRG, Azoic Coupling Component 44
PubChem CID 78170
IUPAC Name N-(4-chloro-2,5-dimethoxyphenyl)-3-oxobutanamide
SMILES COC1=CC(NC(=O)CC(C)=O)=C(OC)C=C1Cl

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 25 g
Formula Weight 271.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-2′,5′-dimethoxyacetoacetanilide
EINECSNumber (3)-0945&(5)-2293
TSCA Yes