accessibility menu, dialog, popup

UserName

CAS RN 4433-79-8

IUPAC Name: N-(4-chloro-2,5-dimethoxyphenyl)-3-oxobutanamide
Synonyms: N-(4-chloro-2,5-dimethoxyphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 271.7
Molecular Formula: C12H14ClNO4
InChi Key: MOUVJGIRLPZEES-UHFFFAOYSA-N
SMILES: COC1=CC(NC(=O)CC(C)=O)=C(OC)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results