missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Chloro-2,2':6',2″-terpyridine 98.0+%, TCI America™
Supplier: TCI America C31871G
Specifications
| 4′-Chloro-2,2′:6′,2′′-terpyridine | |
| 152°C | |
| MFCD00191930 | |
| 4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine | |
| ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 | |
| 267.72 | |
| 267.72 | |
| Crystalline Powder |
| 128143-89-5 | |
| C15H10ClN3 | |
| 1 g | |
| AHEMFMCEBIJRMU-UHFFFAOYSA-N | |
| 4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine | |
| 667748 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 128143-89-5 | |
| 267.72 | |
| AHEMFMCEBIJRMU-UHFFFAOYSA-N | |
| 667748 | |
| ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 |
| C15H10ClN3 | |
| MFCD00191930 | |
| 4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine | |
| 4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine |
Safety and Handling
TSCA : No