missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2,2':6',2″-terpyridine 98.0+%, TCI America™

Catalog No. p-7125880
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3187200MG 200 mg
C31871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3187200MG Supplier TCI America Supplier No. C3187200MG
Only null left

Chemical Identifiers

CAS 128143-89-5
Molecular Formula C15H10ClN3
Molecular Weight (g/mol) 267.72
MDL Number MFCD00191930
InChI Key AHEMFMCEBIJRMU-UHFFFAOYSA-N
Synonym 4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine
PubChem CID 667748
IUPAC Name 4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine
SMILES ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 152°C
Quantity 200 mg
Formula Weight 267.72
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-2,2′:6′,2′′-terpyridine