missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2,2,2-trifluoroacetophenone 97.0+%, TCI America™

Catalog No. C17385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17385G 5 g
C17381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17385G Supplier TCI America Supplier No. C17385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 321-37-9
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00018686
InChI Key DYPQUENOGZXOGE-UHFFFAOYSA-N
PubChem CID 67578
IUPAC Name 1-(4-chlorophenyl)-2,2,2-trifluoroethanone
SMILES C1=CC(=CC=C1C(=O)C(F)(F)F)Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 208.56
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Chloro-2,2,2-trifluoroacetophenone