accessibility menu, dialog, popup

UserName

CAS RN 321-37-9

IUPAC Name: 1-(4-chlorophenyl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(4-chlorophenyl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: DYPQUENOGZXOGE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results