missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,1,3-benzoxadiazole 98.0+%, TCI America™

Catalog No. C13375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13375G 5 g
C133725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13375G Supplier TCI America Supplier No. C13375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7116-16-7
Molecular Formula C6H3ClN2O
Molecular Weight (g/mol) 154.553
MDL Number MFCD00191329
InChI Key YLEBWUGKYMZPQQ-UHFFFAOYSA-N
Synonym 4-Chlorobenzofurazan
PubChem CID 592984
IUPAC Name 4-chloro-2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C(=C1)Cl

Specifications

Melting Point 82°C
Color Yellow
Quantity 5 g
Formula Weight 154.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,1,3-benzoxadiazole