accessibility menu, dialog, popup

UserName

CAS RN 7116-16-7

IUPAC Name: 4-chloro-2,1,3-benzoxadiazole
Synonyms: 4-chloro-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 154.55
Molecular Formula: C6H3ClN2O
InChi Key: YLEBWUGKYMZPQQ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=NON=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results