missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1H-imidazole, 98%

Catalog No. AC460030010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460030010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460030010 Supplier Thermo Scientific Chemicals Supplier No. 460030010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15965-31-8 | C3H3ClN2 | 102.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15965-31-8
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.52
MDL Number MFCD09033087
InChI Key BQRBAXFOPZRMCU-UHFFFAOYSA-N
Synonym 4-chloroimidazole, 4-chloro-1h-imidazole, 4-cl-pyrazole, 5-chloroimidazole, 1h-imidazole, 4-chloro, 1h-imidazole, 5-chloro, 4-chloro-3h-imidazole, 5-chloranyl-1h-imidazole, ksc179g8b
PubChem CID 140019
IUPAC Name 5-chloro-1H-imidazole
SMILES ClC1=CN=CN1

Specifications

Melting Point 118.0°C to 120.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 102.52
Percent Purity 98%
Chemical Name or Material 4-Chloro-1H-imidazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only