missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-naphthol 98.0+%, TCI America™

Catalog No. p-7125387
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22911G 1 g
C22915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22911G Supplier TCI America Supplier No. C22911G
Only null left

Chemical Identifiers

CAS 604-44-4
Molecular Formula C10H7ClO
Molecular Weight (g/mol) 178.615
MDL Number MFCD00003974
InChI Key LVSPDZAGCBEQAV-UHFFFAOYSA-N
Synonym 4-chloro-1-naphthol, 1-chloro-4-hydroxynaphthalene, 1-naphthalenol, 4-chloro, 4-chloro-1-naphthalenol, 1-naphthol, 4-chloro, 4-chloronaphthol, 4-chloro-.alpha.-naphthol, 4-chloro-1-hydroxynaphthalene, 4-chloro-alpha-naphthol, acmc-20amm7
PubChem CID 11787
IUPAC Name 4-chloronaphthalen-1-ol
SMILES C1=CC=C2C(=C1)C(=CC=C2Cl)O

Specifications

Melting Point 120°C
Quantity 1 g
Formula Weight 178.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-1-naphthol [for Biochemical Research]