missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-naphthol, 97%

Catalog No. AC109550050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109550050 5 g Glass bottle
AC109550250 25 g Glass bottle
AC109551000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC109550050 Supplier Thermo Scientific Chemicals Supplier No. 109550050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 604-44-4 | C10H7ClO | 178.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 604-44-4
Molecular Formula C10H7ClO
Molecular Weight (g/mol) 178.62
MDL Number MFCD00003974
InChI Key LVSPDZAGCBEQAV-UHFFFAOYSA-N
Synonym 4-chloro-1-naphthol, 1-chloro-4-hydroxynaphthalene, 1-naphthalenol, 4-chloro, 4-chloro-1-naphthalenol, 1-naphthol, 4-chloro, 4-chloronaphthol, 4-chloro-.alpha.-naphthol, 4-chloro-1-hydroxynaphthalene, 4-chloro-alpha-naphthol, acmc-20amm7
PubChem CID 11787
IUPAC Name 4-chloronaphthalen-1-ol
SMILES C1=CC=C2C(=C1)C(=CC=C2Cl)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 117°C to 120°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC10H6OH
Quantity 5 g
Beilstein 06, 611
Solubility Information Solubility in water: negligible in water. Other solubilities: soluble in methanol and 8% 2,2′oxydiethanol
Formula Weight 178.62
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material 4-Chloro-1-naphthol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-068-5
TSCA TSCA

RUO – Research Use Only