missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-butanol (contains varying amounts of Tetrahydrofuran) 85.0+%, TCI America™

Supplier:  TCI America C0144500G

 View more versions of this product

Catalog No. C0144500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Chloro-1-butanol (contains varying amounts of Tetrahydrofuran)
Yellow
C4H9ClO
500 g
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
OCCCCCl
108.57
CHEBI:87310
≥85.0% (GC)
928-51-8
52°C
MFCD00002967
1993
HXHGULXINZUGJX-UHFFFAOYSA-N
4-chlorobutan-1-ol
13569
108.57
Liquid

Chemical Identifiers

928-51-8
108.57
HXHGULXINZUGJX-UHFFFAOYSA-N
13569
4-chlorobutan-1-ol
C4H9ClO
MFCD00002967
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
CHEBI:87310
OCCCCCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2002

RTECSNumber : EL1325000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions