missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-butanol (contains varying amounts of Tetrahydrofuran) 85.0+%, TCI America™

Catalog No. p-7124245
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
C014425G 25 g
C0144500G 500 g
2 options

Catalog No. C014425G

Supplier: TCI America C014425G

Only null left

Chemical Identifiers

CAS 928-51-8
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00002967
InChI Key HXHGULXINZUGJX-UHFFFAOYSA-N
Synonym 4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
PubChem CID 13569
ChEBI CHEBI:87310
IUPAC Name 4-chlorobutan-1-ol
SMILES OCCCCCl

Specifications

Color Yellow
Boiling Point 52°C
Quantity 25 g
UN Number 1993
Formula Weight 108.57
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-1-butanol (contains varying amounts of Tetrahydrofuran)
EINECSNumber (2)-2002
RTECSNumber EL1325000
TSCA Yes
Recommended Storage Freezer