missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,2-difluorobenzene, 98%

Catalog No. p-7020838
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1292406 5 g
AAA1292414 25 g
AAA1292422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1292406 Supplier Thermo Scientific Chemicals Supplier No. A1292406
Only null left

CAS: 696-02-6 | C6H3ClF2 | 148.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 696-02-6
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042572
InChI Key OPQMRQYYRSTBME-UHFFFAOYSA-N
Synonym 1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
PubChem CID 136519
IUPAC Name 4-chloro-1,2-difluorobenzene
SMILES C1=CC(=C(C=C1Cl)F)F

Specifications

Density 1.35
Boiling Point 128°C to 130°C
Flash Point 32°C (89°F)
Odor Aromatic
Refractive Index 1.4745
Quantity 5 g
UN Number UN1993
Beilstein 2081081
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 4-Chloro-1,2-difluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only