missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxynaphthalene-1-boronic acid, 95%

Catalog No. aah5654203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5654203 1 g
AAH56542MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5654203 Supplier Thermo Scientific Chemicals Supplier No. H5654203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 332398-57-9 | C11H9BO4 | 216.00 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 332398-57-9
Molecular Formula C11H9BO4
Molecular Weight (g/mol) 216.00
MDL Number MFCD06203538
InChI Key SRYYKYNBMCQCAB-UHFFFAOYSA-N
Synonym 4-carboxynaphthalene-1-boronic acid, 4-borono-1-naphthoic acid, 4-dihydroxyboranyl naphthalene-1-carboxylic acid, acmc-209hzn, 4-carboxy-1-naphthylboronic acid, 4-carboxynaphthalen-1-ylboronic acid
PubChem CID 22711863
IUPAC Name 4-borononaphthalene-1-carboxylic acid
SMILES OB(O)C1=C2C=CC=CC2=C(C=C1)C(O)=O

Specifications

Quantity 1 g
Formula Weight 216
Percent Purity 95%
Chemical Name or Material 4-Carboxynaphthalene-1-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only